Assay Details
| Gene Symbol: Ppp2r2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026391.2 | NP_080667.1 | 7 - 8 | 949 | 119 | |
| GenBank mRNA | AF366393.1 | - | 7 - 8 | 857 | 119 | |
| AK005033.1 | - | 6 - 7 | 838 | 119 | ||
| AK011198.1 | - | 7 - 8 | 969 | 119 | ||
| AK129386.2 | - | 7 - 8 | 976 | 119 | ||
| BC066022.1 | - | 7 - 8 | 949 | 6828707 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r2d
Genomic Map