Assay Details
| Gene Symbol: Knstrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026412.3 | NP_080688.2 | 5 - 6 | 680 | 65 | |
| GenBank mRNA | AK006328.1 | - | 5 - 6 | 681 | 65 | |
| AK012148.1 | - | 5 - 6 | 480 | 65 | ||
| AK013184.1 | - | 5 - 6 | 428 | 65 | ||
| AK028590.1 | - | 5 - 6 | 481 | 65 | ||
| AK036374.1 | - | 5 - 6 | 469 | 65 | ||
| BC031709.1 | - | 4 - 5 | 388 | 4021550 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Knstrn
Genomic Map