Assay Details
| Gene Symbol: Dcxr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026428.2 | NP_080704.2 | 1 - 2 | 104 | 108 | |
| GenBank mRNA | AK004023.1 | - | 1 - 2 | 103 | 108 | |
| AK007627.1 | - | 1 - 2 | 116 | 108 | ||
| AK153521.1 | - | 1 - 2 | 85 | 108 | ||
| BC012247.1 | - | 1 - 2 | 93 | 4189635 | 108 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK004023.1,AK007627.1,AK153521.1,BC012247.1,NM_026428.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
xylulose metabolic process
glucose metabolic process
NADP metabolic process
metabolic process
D-xylose metabolic process
protein homotetramerization
oxidation-reduction process
protein binding
oxidoreductase activity
oxidoreductase activity, acting on CH-OH group of donors
oxidoreductase activity, acting on NAD(P)H, quinone or similar compound as acceptor
L-xylulose reductase (NADP+) activity
carbohydrate metabolic process
xylulose metabolic process
glucose metabolic process
NADP metabolic process
metabolic process
D-xylose metabolic process
protein homotetramerization
oxidation-reduction process
protein binding
oxidoreductase activity
oxidoreductase activity, acting on CH-OH group of donors
oxidoreductase activity, acting on NAD(P)H, quinone or similar compound as acceptor
L-xylulose reductase (NADP+) activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28186434 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28186436 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28186435 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P181319 | AK161732.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P181319 | AK153521.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P181319 | D89656.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P181319 | NM_026428.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P181319 | AK004023.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P181319 | AK161732.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P181319 | AK153521.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P181319 | D89656.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P181319 | NM_026428.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P181319 | AK004023.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 852747 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 289578 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 39890 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167446 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P181319 | AK161732.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P181319 | AK153521.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P181319 | D89656.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P181319 | NM_026428.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P181319 | AK004023.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P181319 | AK161732.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P181319 | AK153521.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P181319 | D89656.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P181319 | NM_026428.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P181319 | AK004023.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P181312 | AK161732.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P181312 | AK153521.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P181312 | D89656.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P181312 | NM_026428.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P181312 | AK002803.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P181319 | AK161732.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P181319 | AK153521.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P181319 | D89656.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P181319 | NM_026428.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P181319 | AK004023.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P181319 | AK161732.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P181319 | AK153521.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P181319 | D89656.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P181319 | NM_026428.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P181319 | AK004023.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28186448 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167446 | - | - |
| Whole Mouse Genome G4122A | A_52_P98100 | AK161732.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181312 | AK161732.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181312 | AK153521.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181312 | D89656.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181312 | NM_026428.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181312 | AK002803.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181319 | AK161732.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181319 | AK153521.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181319 | D89656.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181319 | NM_026428.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181319 | AK004023.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181319 | AK161732.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181319 | AK153521.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181319 | D89656.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181319 | NM_026428.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181319 | AK004023.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102496 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167446 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09620636 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09540886 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09620650 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09620623 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09540888 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28186435 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28186436 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28186434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28186422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09620651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09620632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09383014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09620623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09227461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09620636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09620633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28186426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09620647 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10393928 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393927 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393933 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393929 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393931 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393930 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393934 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393932 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393935 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419456_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10393926 | - | - |
| Mouse Genome 430 2.0 Array | 1419456_at | - | - |
| Mouse Genome 430 2.0 Array | 1419456_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67880_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67880_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100040457.mm.1 | NM_026428.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040457.mm.1 | NM_026428.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040457.mm.1 | TC1100004228.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100040458.mm.1 | NM_026428.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040458.mm.1 | NM_026428.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040458.mm.1 | TC1100004228.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100040461.mm.1 | NM_026428.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040461.mm.1 | NM_026428.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040461.mm.1 | TC1100004228.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419456_at | - | - |
| Mouse Expression Set 430 | 1419456_PM_at | - | - |
| Murine Genome U74v2 Set | 165948_i_at | - | - |
| Murine Genome U74v2 Set | 165948_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419456_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419456_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419456_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419456_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4730551 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004730551 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2759109 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2759109 | - | - |
| mouse-6 v1.1 (Illumina) | scl39223.3.102_51-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4730551 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39223.3.102_51-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4730551 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2868280 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2759108 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2868280 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2759108 | - | - |

Dcxr
Genomic Map