Assay Details
| Gene Symbol: Coq5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026504.2 | NP_080780.1 | 3 - 4 | 590 | 80 | |
| GenBank mRNA | AK007496.1 | - | 3 - 4 | 591 | 80 | |
| AK009825.1 | - | 3 - 4 | 586 | 80 | ||
| AK014348.1 | - | 3 - 4 | 602 | 80 | ||
| AK045993.1 | - | 3 - 4 | 597 | 80 | ||
| AK050334.1 | - | 3 - 4 | 597 | 80 | ||
| AK196745.1 | - | 1 - 2 | 199 | 80 | ||
| BC052345.1 | - | 3 - 4 | 597 | 6585618 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Coq5
Genomic Map