Assay Details
| Gene Symbol: Coq5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026504.2 | NP_080780.1 | 4 - 5 | 688 | 54 | |
| GenBank mRNA | AK007496.1 | - | 4 - 5 | 689 | 54 | |
| AK009825.1 | - | 4 - 5 | 684 | 54 | ||
| AK014348.1 | - | 4 - 5 | 700 | 54 | ||
| AK045993.1 | - | 4 - 5 | 695 | 54 | ||
| AK050334.1 | - | 4 - 5 | 695 | 54 | ||
| BC052345.1 | - | 4 - 5 | 695 | 6585618 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Coq5
Genomic Map