Assay Details
| Gene Symbol: Ddx56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026538.3 | NP_080814.1 | 3 - 4 | 389 | 66 | |
| GenBank mRNA | AK011136.1 | - | 3 - 4 | 399 | 66 | |
| AK076047.1 | - | 3 - 4 | 407 | 66 | ||
| AK076975.1 | - | 3 - 4 | 389 | 66 | ||
| AK077514.1 | - | 3 - 4 | 402 | 66 | ||
| AK145628.1 | - | 3 - 4 | 430 | 66 | ||
| AK159793.1 | - | 3 - 4 | 393 | 66 | ||
| AK169654.1 | - | 3 - 4 | 404 | 66 | ||
| BC018291.1 | - | 3 - 4 | 383 | 4190167 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx56
Genomic Map