Assay Details
| Gene Symbol: Nt5c3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102650.1 | NP_001096120.1 | 2 - 3 | 145 | 86 | |
| NM_026561.4 | NP_080837.3 | 2 - 3 | 145 | 86 | ||
| GenBank mRNA | AK006972.1 | - | 3 - 4 | 354 | 86 | |
| AK082816.1 | - | 2 - 3 | 131 | 86 | ||
| AK148222.1 | - | 2 - 3 | 121 | 86 | ||
| AK162770.1 | - | 1 - 2 | 223 | 86 | ||
| AK166393.1 | - | 2 - 3 | 121 | 86 | ||
| BC015307.1 | - | 1 - 2 | 100 | 4239117 | 86 | |
| BY047327.1 | - | 2 - 3 | 145 | 86 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK006972.1,AK082816.1,AK162770.1,BC015307.1 transcripts.
Set Membership: |
|

Nt5c3b
Genomic Map