Assay Details
| Gene Symbol: Rhpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027897.4 | NP_082173.3 | 4 - 5 | 492 | 103 | |
| GenBank mRNA | AK004849.1 | - | 4 - 5 | 415 | 103 | |
| AK050214.1 | - | 4 - 5 | 406 | 103 | ||
| BC100314.1 | - | 4 - 5 | 399 | 30848276 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhpn2
Genomic Map