Assay Details
| Gene Symbol: Rhpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027897.4 | NP_082173.3 | 8 - 9 | 1059 | 95 | |
| GenBank mRNA | AK004849.1 | - | 8 - 9 | 982 | 95 | |
| AK050214.1 | - | 8 - 9 | 973 | 95 | ||
| BC100314.1 | - | 8 - 9 | 966 | 30848276 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhpn2
Genomic Map