Assay Details
| Gene Symbol: Cnot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037846.3 | NP_001032935.2 | 1 - 2 | 54 | 50 | |
| NM_001037848.3 | NP_001032937.1 | 1 - 2 | 54 | 50 | ||
| NM_028082.2 | NP_082358.2 | 1 - 2 | 54 | 50 | ||
| NR_024327.1 | 1 - 2 | 54 | 50 | |||
| GenBank mRNA | AK011231.1 | - | 1 - 2 | 44 | 50 | |
| AK078121.1 | - | 1 - 2 | 44 | 50 | ||
| AK149767.1 | - | 1 - 2 | 28 | 50 | ||
| AK167834.1 | - | 1 - 2 | 32 | 50 | ||
| AK172132.1 | - | 1 - 2 | 41 | 50 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK011231.1,AK078121.1,AK149767.1,AK167834.1,NM_001037846.3,NM_001037848.3,NM_028082.2 transcripts.
Set Membership: |
|

Cnot2
Genomic Map