Assay Details
| Gene Symbol: Brap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289544.1 | NP_001276473.1 | 3 - 4 | 768 | 74 | |
| NM_028227.3 | NP_082503.2 | 3 - 4 | 640 | 74 | ||
| GenBank mRNA | AF321920.1 | - | 3 - 4 | 532 | 74 | |
| AF321921.1 | - | 3 - 4 | 769 | 74 | ||
| AK013885.1 | - | 1 - 2 | 133 | 74 | ||
| BC062932.1 | - | 3 - 4 | 616 | 74 | ||
| BF467152.1 | - | 3 - 4 | 553 | 74 | ||
| CD355617.1 | - | 3 - 4 | 616 | 74 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK013885.1 transcripts.
Set Membership: |
|

Brap
Genomic Map