Assay Details
| Gene Symbol: Cul4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110142.1 | NP_001103612.1 | 3 - 4 | 362 | 72 | |
| NM_028288.5 | NP_082564.3 | 3 - 4 | 303 | 72 | ||
| GenBank mRNA | AK012410.1 | - | 1 - 2 | 66 | 72 | |
| AK032701.1 | - | 3 - 4 | 304 | 72 | ||
| AK160998.1 | - | 1 - 2 | 73 | 72 | ||
| AY330868.1 | - | 3 - 4 | 304 | 72 | ||
| BC004026.1 | - | 3 - 4 | 351 | 3494708 | 72 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC004026.1 transcripts.
Set Membership: |
|

Cul4b
Genomic Map