Assay Details
| Gene Symbol: Cul4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110142.1 | NP_001103612.1 | 20 - 21 | 2616 | 113 | |
| NM_028288.5 | NP_082564.3 | 20 - 21 | 2557 | 113 | ||
| GenBank mRNA | AB093259.1 | - | 14 - 15 | 4676 | 113 | |
| AK012410.1 | - | 18 - 19 | 2320 | 113 | ||
| AK032701.1 | - | 20 - 21 | 2558 | 113 | ||
| AK160998.1 | - | 18 - 19 | 2327 | 113 | ||
| AK170632.1 | - | 16 - 17 | 2614 | 113 | ||
| AY330868.1 | - | 20 - 21 | 2558 | 113 | ||
| BC004026.1 | - | 20 - 21 | 2605 | 3494708 | 113 | |
| BC010347.1 | - | 7 - 8 | 743 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cul4b
Genomic Map