Assay Details
| Gene Symbol: Arhgap10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030113.2 | NP_084389.2 | 2 - 3 | 369 | 96 | |
| GenBank mRNA | AF297030.1 | - | 2 - 3 | 332 | 96 | |
| AK020920.1 | - | 2 - 3 | 370 | 96 | ||
| AK041867.1 | - | 2 - 3 | 416 | 96 | ||
| AY179966.1 | - | 2 - 3 | 245 | 96 | ||
| AY179967.1 | - | 2 - 3 | 245 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap10
Genomic Map