Assay Details
| Gene Symbol: Arhgap10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030113.2 | NP_084389.2 | 14 - 15 | 1426 | 69 | |
| GenBank mRNA | AF297029.1 | - | 14 - 15 | 1330 | 69 | |
| AF297030.1 | - | 14 - 15 | 1389 | 69 | ||
| AY179965.1 | - | 14 - 15 | 1302 | 69 | ||
| AY179966.1 | - | 13 - 14 | 1236 | 69 | ||
| AY179967.1 | - | 14 - 15 | 1302 | 69 | ||
| BU151977.1 | - | 4 - 5 | 252 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap10
Genomic Map