Assay Details
| Gene Symbol: Parp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030253.3 | NP_084529.1 | 2 - 3 | 700 | 73 | |
| GenBank mRNA | AK050032.1 | - | 2 - 3 | 144 | 73 | |
| BC003281.1 | - | 2 - 3 | 142 | 3581841 | 73 | |
| BC070466.1 | - | 2 - 3 | 700 | 30615225 | 73 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK050032.1,BC003281.1,BC070466.1,NM_030253.3 transcripts.
Set Membership: |
|

Parp9
Genomic Map