Assay Details
| Gene Symbol: Zxdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030260.3 | NP_084536.2 | 3 - 4 | 1304 | 67 | |
| NM_173002.3 | NP_766590.1 | 3 - 4 | 1304 | 67 | ||
| GenBank mRNA | AK030287.1 | - | 3 - 4 | 1220 | 67 | |
| AK042190.1 | - | 3 - 4 | 1231 | 67 | ||
| AK042392.1 | - | 3 - 4 | 1214 | 67 | ||
| AK044429.1 | - | 3 - 4 | 1197 | 67 | ||
| AK044529.1 | - | 3 - 4 | 1228 | 67 | ||
| AK044948.1 | - | 3 - 4 | 1198 | 67 | ||
| AK047079.1 | - | 3 - 4 | 1186 | 67 | ||
| AK047538.1 | - | 3 - 4 | 1220 | 67 | ||
| AK080307.1 | - | 3 - 4 | 1198 | 67 | ||
| AK081005.1 | - | 3 - 4 | 1306 | 67 | ||
| AK169517.1 | - | 3 - 4 | 1191 | 67 | ||
| BC003332.1 | - | 3 - 4 | 1288 | 3256958 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zxdc
Genomic Map