Assay Details
| Gene Symbol: Pofut2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030262.3 | NP_084538.2 | 3 - 4 | 550 | 62 | |
| GenBank mRNA | AF455270.1 | - | 3 - 4 | 540 | 62 | |
| AK132373.1 | - | 3 - 4 | 547 | 62 | ||
| AK148244.1 | - | 3 - 4 | 549 | 62 | ||
| AK190890.1 | - | 1 - 2 | 114 | 62 | ||
| BC018194.1 | - | 3 - 4 | 548 | 62 | ||
| BC147077.1 | - | 3 - 4 | 617 | 9056064 | 62 | |
| BC147078.1 | - | 3 - 4 | 617 | 9056063 | 62 | |
| CO814561.1 | - | 550 | 62 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Pofut2
Genomic Map