Assay Details
| Gene Symbol: Pofut2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030262.3 | NP_084538.2 | 7 - 8 | 1030 | 59 | |
| GenBank mRNA | AF455270.1 | - | 7 - 8 | 1020 | 59 | |
| AK132373.1 | - | 7 - 8 | 1027 | 59 | ||
| AK147414.1 | - | 3 - 4 | 5838 | 59 | ||
| AK148244.1 | - | 7 - 8 | 1030 | 59 | ||
| BC003494.1 | - | 3 - 4 | 382 | 59 | ||
| BC018194.1 | - | 7 - 8 | 1028 | 59 | ||
| BC147077.1 | - | 7 - 8 | 1097 | 9056064 | 59 | |
| BC147078.1 | - | 7 - 8 | 1097 | 9056063 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pofut2
Genomic Map