Assay Details
| Gene Symbol: Rabep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030566.2 | NP_085043.2 | 4 - 5 | 869 | 75 | |
| GenBank mRNA | AK029188.1 | - | 2 - 3 | 227 | 75 | |
| AK089938.1 | - | 4 - 5 | 573 | 75 | ||
| AK144836.1 | - | 4 - 5 | 555 | 75 | ||
| AK157832.1 | - | 4 - 5 | 530 | 75 | ||
| BC004088.1 | - | 3 - 4 | 615 | 3593722 | 75 | |
| BC015287.1 | - | 4 - 5 | 877 | 4240442 | 75 | |
More Information
Set Membership: |
|

Rabep2
Genomic Map