Assay Details
| Gene Symbol: Rabep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030566.2 | NP_085043.2 | 8 - 9 | 1569 | 63 | |
| GenBank mRNA | AK029188.1 | - | 6 - 7 | 927 | 63 | |
| AK144836.1 | - | 8 - 9 | 1255 | 63 | ||
| AK157832.1 | - | 8 - 9 | 1230 | 63 | ||
| BC004088.1 | - | 7 - 8 | 1315 | 3593722 | 63 | |
| BC015287.1 | - | 8 - 9 | 1577 | 4240442 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rabep2
Genomic Map