Assay Details
| Gene Symbol: Rabep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030566.2 | NP_085043.2 | 12 - 13 | 1923 | 88 | |
| GenBank mRNA | AK029188.1 | - | 9 - 10 | 1225 | 88 | |
| AK089938.1 | - | 12 - 13 | 1629 | 88 | ||
| AK134248.1 | - | 132 | 88 | |||
| AK144836.1 | - | 12 - 13 | 1609 | 88 | ||
| AK157832.1 | - | 12 - 13 | 1584 | 88 | ||
| BC004088.1 | - | 11 - 12 | 1669 | 3593722 | 88 | |
| BC015287.1 | - | 11 - 12 | 2172 | 4240442 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25500413 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25500407 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25500411 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25500409 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25500412 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2053958 | BC004088.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2053958 | BC015287.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2053958 | BY665800.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2053958 | AK144836.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2053958 | AK157832.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816527 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 444524 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 690526 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 476853 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 402305 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 69055 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972544 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494150 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 657762 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676517 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 398050 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 115928 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16180 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125305 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166198 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053958 | BC004088.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053958 | BC015287.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053958 | BY665800.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053958 | AK144836.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053958 | AK157832.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500421 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06358882 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01629734 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125305 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166198 | - | - |
| Whole Mouse Genome G4122A | A_51_P484959 | BC004088.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P484959 | BC015287.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P484959 | BY665800.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P484959 | AK144836.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P484959 | AK157832.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126165 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166198 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125305 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06429636 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06429614 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06429631 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06429629 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06429617 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06429630 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25500407 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25500409 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25500412 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25500411 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25500413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06654222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06517425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06429630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06249284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06429633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06423660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06465267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06413524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06429634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06251266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500442 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10557419 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557408 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557406 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557411 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557413 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557409 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557418 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557412 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557415 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557410 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557414 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557416 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440795_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425602_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10557405 | - | - |
| Mouse Genome 430 2.0 Array | 1425602_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1440795_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1440795_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1425602_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70314_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70314_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700015363.mm.1 | TC0700001802.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700015363.mm.1 | NM_030566.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015363.mm.1 | NM_030566.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015364.mm.1 | TC0700001802.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700015364.mm.1 | NM_030566.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700015364.mm.1 | NM_030566.2 | Refseq |
| Mouse Expression Set 430 | 1440795_x_at | - | - |
| Mouse Expression Set 430 | 1440795_PM_x_at | - | - |
| Murine Genome U74v2 Set | 165714_PM_f_at | - | - |
| Murine Genome U74v2 Set | 165714_f_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425602_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425602_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440795_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440795_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6130673 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006130673 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2599751 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2599751 | - | - |
| mouse-6 v1.1 (Illumina) | scl32063.14.1_155-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6130673 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32063.14.1_155-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6130673 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2599751 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2599751 | - | - |

Rabep2
Genomic Map