Assay Details
| Gene Symbol: Kcnip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276358.1 | NP_001263287.1 | 3 - 4 | 290 | 100 | |
| NM_030716.3 | NP_109641.2 | 4 - 5 | 625 | 100 | ||
| NM_145703.2 | NP_663749.1 | 5 - 6 | 679 | 100 | ||
| NM_145704.2 | NP_663750.1 | 3 - 4 | 529 | 100 | ||
| GenBank mRNA | AF439339.1 | - | 5 - 6 | 419 | 100 | |
| AF439340.1 | - | 4 - 5 | 365 | 100 | ||
| AF439341.1 | - | 3 - 4 | 269 | 100 | ||
| AJ278535.1 | - | 4 - 5 | 215 | 100 | ||
| AJ278536.1 | - | 5 - 6 | 365 | 100 | ||
| AJ278537.1 | - | 4 - 5 | 311 | 100 | ||
| AY647241.1 | - | 3 - 4 | 290 | 100 | ||
| BC056434.1 | - | 4 - 5 | 1396 | 100 | ||
| BC132215.1 | - | 2 - 3 | 303 | 40130492 | 100 | |
| BC138496.1 | - | 3 - 4 | 345 | 8861516 | 100 | |
| DQ148496.1 | - | 5 - 6 | 419 | 100 | ||
| DQ148497.1 | - | 4 - 5 | 365 | 100 | ||
| DQ148498.1 | - | 3 - 4 | 269 | 100 | ||
| DQ148505.1 | - | 3 - 4 | 287 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnip2
Genomic Map