Assay Details
| Gene Symbol: Crispld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031402.2 | NP_113579.2 | 2 - 3 | 667 | 75 | |
| GenBank mRNA | AF329198.1 | - | 2 - 3 | 664 | 75 | |
| AK132405.1 | - | 2 - 3 | 405 | 75 | ||
| AK133870.1 | - | 2 - 3 | 668 | 75 | ||
| AK134273.1 | - | 2 - 3 | 603 | 75 | ||
| AK137611.1 | - | 2 - 3 | 668 | 75 | ||
| BC040768.1 | - | 2 - 3 | 996 | 4458200 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Crispld1
Genomic Map