Assay Details
| Gene Symbol: Gpr87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF295366.1 | - | 1 | 56 | 86 | |
| BC125653.1 | - | 1 | 85 | 40130168 | 86 | |
| BC125655.1 | - | 1 | 85 | 40130170 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpr87
Genomic Map