Assay Details
| Gene Symbol: Dxo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163770.1 | NP_001157242.1 | 3 - 4 | 794 | 89 | |
| NM_033613.2 | NP_291091.2 | 3 - 4 | 819 | 89 | ||
| GenBank mRNA | BC004713.1 | - | 3 - 4 | 755 | 3499698 | 89 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
mRNA catabolic process
metabolic process
RNA destabilization
nuclear mRNA surveillance
nucleic acid phosphodiester bond hydrolysis
nucleotide binding
3'-5'-exoribonuclease activity
magnesium ion binding
RNA binding
mRNA binding
nuclease activity
exonuclease activity
5'-3' exoribonuclease activity
thiamine-pyrophosphatase activity
5'-3' exonuclease activity
8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity
UDP-2,3-diacylglucosamine hydrolase activity
bis(5'-nucleosyl)-tetraphosphatase activity
dATP pyrophosphohydrolase activity
pyrophosphatase activity
hydrolase activity
dihydroneopterin monophosphate phosphatase activity
dihydroneopterin triphosphate pyrophosphohydrolase activity
RNA pyrophosphohydrolase activity
dITP diphosphatase activity
dTTP diphosphatase activity
XTP diphosphatase activity
ATP-dependent 5'-3' DNA helicase activity
phosphocholine hydrolase activity
metal ion binding
mRNA catabolic process
metabolic process
RNA destabilization
nuclear mRNA surveillance
nucleic acid phosphodiester bond hydrolysis
nucleotide binding
3'-5'-exoribonuclease activity
magnesium ion binding
RNA binding
mRNA binding
nuclease activity
exonuclease activity
5'-3' exoribonuclease activity
thiamine-pyrophosphatase activity
5'-3' exonuclease activity
8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity
UDP-2,3-diacylglucosamine hydrolase activity
bis(5'-nucleosyl)-tetraphosphatase activity
dATP pyrophosphohydrolase activity
pyrophosphatase activity
hydrolase activity
dihydroneopterin monophosphate phosphatase activity
dihydroneopterin triphosphate pyrophosphohydrolase activity
RNA pyrophosphohydrolase activity
dITP diphosphatase activity
dTTP diphosphatase activity
XTP diphosphatase activity
ATP-dependent 5'-3' DNA helicase activity
phosphocholine hydrolase activity
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156012 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156011 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156013 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156010 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156009 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156014 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156008 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267388 | BC004713.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267388 | NM_001163770.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267388 | NM_033613.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267388 | BC004713.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267388 | NM_001163770.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267388 | NM_033613.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499805 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125912 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267388 | BC004713.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267388 | NM_001163770.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267388 | NM_033613.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267388 | BC004713.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267388 | NM_001163770.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267388 | NM_033613.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267388 | BC004713.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267388 | NM_001163770.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267388 | NM_033613.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267388 | BC004713.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267388 | NM_001163770.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267388 | NM_033613.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156016 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125912 | - | - |
| Whole Mouse Genome G4122A | A_51_P267388 | BC004713.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267388 | NM_001163770.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267388 | NM_033613.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267388 | BC004713.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267388 | NM_001163770.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267388 | NM_033613.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112868 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138083 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125912 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13174868 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13174878 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13174879 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13174886 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156013 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156012 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156009 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156008 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156014 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156010 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13174879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13174883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13174877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156008 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10444503 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444502 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444497 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444499 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444501 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444498 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444504 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423116_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10444496 | - | - |
| Mouse Genome 430 2.0 Array | 1423116_at | - | - |
| Mouse Genome 430 2.0 Array | 1423116_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 112403_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 112403_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700006380.mm.1 | NM_001163770.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006380.mm.1 | NM_001163770.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006380.mm.1 | NM_033613.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006380.mm.1 | NM_033613.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006380.mm.1 | TC1700000646.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700006379.mm.1 | NM_001163770.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006379.mm.1 | NM_001163770.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006379.mm.1 | NM_033613.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006379.mm.1 | NM_033613.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006379.mm.1 | TC1700000646.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700006377.mm.1 | NM_001163770.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006377.mm.1 | NM_001163770.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006377.mm.1 | NM_033613.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006377.mm.1 | NM_033613.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006377.mm.1 | TC1700000646.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423116_at | - | - |
| Mouse Expression Set 430 | 1423116_PM_at | - | - |
| Murine Genome U74v2 Set | 97428_PM_at | - | - |
| Murine Genome U74v2 Set | 97428_at | - | - |
| Murine Genome U74v2 Set | 171165_PM_r_at | - | - |
| Murine Genome U74v2 Set | 171165_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423116_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423116_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423116_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423116_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 447256 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2360068 | - | - |
| MouseWG-6 v2 Expression BeadChip | 3870358 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2360068 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3870358 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002360068 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003870358 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2694156 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2653508 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2653508 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2694156 | - | - |
| mouse-6 v1.1 (Illumina) | scl50807.7.1_59-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0001747.1_14-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2360068 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3870358 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001747.1_14-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50807.7.1_59-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2360068 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3870358 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2694156 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1233838 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2694156 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1233838 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl50807.7.1_59-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001747.1_14-S | - | - |

Dxo
Genomic Map