Assay Details
| Gene Symbol: Dhcr24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053272.2 | NP_444502.2 | 2 - 3 | 509 | 122 | |
| GenBank mRNA | AK017937.1 | - | 2 - 3 | 271 | 122 | |
| AK083737.1 | - | 2 - 3 | 464 | 122 | ||
| AK129036.1 | - | 2 - 3 | 509 | 122 | ||
| AY039762.1 | - | 2 - 3 | 397 | 122 | ||
| BC019797.1 | - | 2 - 3 | 453 | 5054108 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhcr24
Genomic Map