Assay Details
| Gene Symbol: Ftcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080845.2 | NP_543121.1 | 1 - 2 | 110 | 70 | |
| GenBank mRNA | AK165414.1 | - | 1 - 2 | 108 | 70 | |
| BB610765.1 | - | 111 | 70 | |||
| BC010813.1 | - | 1 - 2 | 72 | 4211065 | 70 | |
| BC024078.1 | - | 1 - 2 | 104 | 5098529 | 70 | |
| CD297973.1 | - | 1 - 2 | 113 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ftcd
Genomic Map