Assay Details
| Gene Symbol: Ftcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080845.2 | NP_543121.1 | 4 - 5 | 516 | 78 | |
| GenBank mRNA | BB610765.1 | - | 517 | 78 | ||
| BC010813.1 | - | 4 - 5 | 478 | 4211065 | 78 | |
| BC024078.1 | - | 4 - 5 | 510 | 5098529 | 78 | |
| CD297973.1 | - | 4 - 5 | 519 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ftcd
Genomic Map