Assay Details
| Gene Symbol: Capn8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130890.2 | NP_570960.2 | 16 - 17 | 1819 | 102 | |
| GenBank mRNA | AB050202.1 | - | 16 - 17 | 1815 | 102 | |
| AB061518.1 | - | 16 - 17 | 1765 | 102 | ||
| BC125504.1 | - | 16 - 17 | 1791 | 40130019 | 102 | |
| BC125508.1 | - | 16 - 17 | 1791 | 40130023 | 102 | |
| BC145055.1 | - | 16 - 17 | 1791 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Capn8
Genomic Map