Assay Details
Gene Symbol: Cacng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133190.1 | NP_573453.1 | 3 - 4 | 610 | 66 | |
GenBank mRNA | AF361350.1 | - | 3 - 4 | 610 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transport
ion transport
calcium ion transport
regulation of ion transmembrane transport
calcium ion transmembrane transport
regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
voltage-gated ion channel activity
calcium channel activity
transport
ion transport
calcium ion transport
regulation of ion transmembrane transport
calcium ion transmembrane transport
regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
voltage-gated ion channel activity
calcium channel activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P133409 | NM_133190.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P133409 | AF361350.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 386301 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 207026 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 449182 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906952 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 1147 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 148237 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 657924 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481562 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 603398 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342311 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 254653 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 522888 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773935 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 160676 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 31980 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 152143 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 92242 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P133409 | NM_133190.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P133409 | AF361350.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160935 | NM_133190.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P160935 | AF361350.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P160935 | NM_133190.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P160935 | AF361350.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133409 | NM_133190.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133409 | AF361350.1 | mouse_transcriptome |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017347 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017336 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017321 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017350 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017338 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017322 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017323 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017360 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017331 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017326 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017344 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017320 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017342 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017368 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017370 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017357 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017365 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017352 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017319 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017345 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017334 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017361 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017363 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017354 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017349 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017339 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017351 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017329 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017355 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017328 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017333 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017316 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017366 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017315 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P33017318 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10569705 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10569704 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10569703 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10569706 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451864_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1459579_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10569702 | - | - |
Mouse Genome 430 2.0 Array | 1459579_at | - | - |
Mouse Genome 430 2.0 Array | 1459579_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1451864_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1451864_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700000083.mm.1 | TC0700000009.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700000083.mm.1 | NM_133190.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700000085.mm.1 | TC0700000009.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700000085.mm.1 | NM_133190.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700000084.mm.1 | TC0700000009.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700000084.mm.1 | NM_133190.2 | Refseq |
Mouse Expression Set 430 | 1451864_at | - | - |
Mouse Expression Set 430 | 1451864_PM_at | - | - |
Mouse Expression Set 430 | 1459579_at | - | - |
Mouse Expression Set 430 | 1459579_PM_at | - | - |
Murine Genome U74v2 Set | 137323_at | - | - |
Murine Genome U74v2 Set | 137323_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1451864_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1451864_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451864_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451864_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1459579_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1459579_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 662135 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 610152 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 610152 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000610152 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1230074 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1230074 | - | - |
mouse-6 v1.1 (Illumina) | scl081905.4_2-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 610152 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl081905.4_2-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 610152 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2709881 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2709881 | - | - |
