Assay Details
| Gene Symbol: Slc8b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177594.1 | NP_001171065.1 | 5 - 6 | 623 | 86 | |
| NM_001177595.1 | NP_001171066.1 | 6 - 7 | 674 | 86 | ||
| NM_133221.2 | NP_573484.2 | 6 - 7 | 674 | 86 | ||
| GenBank mRNA | AF261233.1 | - | 6 - 7 | 674 | 86 | |
| AK028537.1 | - | 6 - 7 | 619 | 86 | ||
| AK157173.1 | - | 5 - 6 | 581 | 86 | ||
| BC043689.1 | - | 6 - 7 | 614 | 5137181 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc8b1
Genomic Map