Assay Details
| Gene Symbol: Crb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133239.2 | NP_573502.2 | 7 - 8 | 2838 | 94 | |
| GenBank mRNA | AF406641.1 | - | 7 - 8 | 2803 | 94 | |
| BC053331.1 | - | 2 - 3 | 1081 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crb1
Genomic Map