Assay Details
| Gene Symbol: Cd34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111059.1 | NP_001104529.1 | 4 - 5 | 732 | 99 | |
| NM_133654.3 | NP_598415.1 | 4 - 5 | 732 | 99 | ||
| GenBank mRNA | AK044121.1 | - | 4 - 5 | 668 | 99 | |
| AK089010.1 | - | 4 - 5 | 638 | 99 | ||
| AK167329.1 | - | 4 - 5 | 643 | 99 | ||
| AK167350.1 | - | 4 - 5 | 643 | 99 | ||
| BC006607.1 | - | 4 - 5 | 648 | 3966337 | 99 | |
| BC066820.1 | - | 4 - 5 | 605 | 99 | ||
| BU058977.1 | - | 2 - 3 | 215 | 99 | ||
| S69293.1 | - | 4 - 5 | 610 | 99 | ||
| S69299.1 | - | 4 - 5 | 587 | 99 | ||
| S69301.1 | - | 4 - 5 | 587 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd34
Genomic Map