Assay Details
| Gene Symbol: Itga9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113514.1 | NP_001106986.1 | 7 - 8 | 727 | 68 | |
| NM_133721.2 | NP_598482.2 | 21 - 22 | 2548 | 68 | ||
| GenBank mRNA | AJ344342.1 | - | 21 - 22 | 2551 | 68 | |
| AK083987.1 | - | 6 - 7 | 567 | 68 | ||
| AK139690.1 | - | 7 - 8 | 715 | 68 | ||
| AK153585.1 | - | 19 - 20 | 2010 | 68 | ||
| BC049939.1 | - | 4 - 5 | 344 | 68 | ||
| BC150824.1 | - | 21 - 22 | 2454 | 9087735 | 68 | |
More Information
Set Membership: |
|

Itga9
Genomic Map