Assay Details
| Gene Symbol: Wwc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133791.4 | NP_598552.2 | 18 - 19 | 3223 | 66 | |
| GenBank mRNA | AB045323.1 | - | 18 - 19 | 3248 | 66 | |
| AK028956.1 | - | 840 | 66 | |||
| AK032989.1 | - | 14 - 15 | 2289 | 66 | ||
| AK079375.1 | - | 18 - 19 | 3059 | 66 | ||
| AK147648.1 | - | 3217 | 66 | |||
| BC054434.1 | - | 8 - 9 | 1504 | 66 | ||
| BC067050.1 | - | 18 - 19 | 3223 | 30537299 | 66 | |
More Information
Set Membership: |
|

Wwc2
Genomic Map