Assay Details
| Gene Symbol: Camk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133926.2 | NP_598687.1 | 6 - 7 | 675 | 89 | |
| GenBank mRNA | AK089742.1 | - | 6 - 7 | 658 | 89 | |
| AK134936.1 | - | 6 - 7 | 695 | 89 | ||
| AK170188.1 | - | 5 - 6 | 741 | 89 | ||
| AK208371.1 | - | 1 - 2 | 157 | 89 | ||
| BC014825.1 | - | 6 - 7 | 675 | 3969343 | 89 | |
| BC042494.1 | - | 2 - 3 | 1934 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Camk1
Genomic Map