Assay Details
| Gene Symbol: Camk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133926.2 | NP_598687.1 | 9 - 10 | 944 | 92 | |
| GenBank mRNA | AK089742.1 | - | 9 - 10 | 927 | 92 | |
| AK134936.1 | - | 9 - 10 | 964 | 92 | ||
| AK170188.1 | - | 8 - 9 | 1010 | 92 | ||
| AK188088.1 | - | 1 - 2 | 44 | 92 | ||
| AK217859.1 | - | 1 - 2 | 44 | 92 | ||
| BC014825.1 | - | 9 - 10 | 944 | 3969343 | 92 | |
| BC025510.1 | - | 1 - 2 | 295 | 92 | ||
| BC042494.1 | - | 5 - 6 | 2724 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Camk1
Genomic Map