Assay Details
| Gene Symbol: Gstt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133994.3 | NP_598755.1 | 2 - 3 | 659 | 85 | |
| GenBank mRNA | AF508157.1 | - | 2 - 3 | 659 | 85 | |
| AK138669.1 | - | 2 - 3 | 316 | 85 | ||
| AK154161.1 | - | 2 - 3 | 365 | 85 | ||
| AK168226.1 | - | 2 - 3 | 308 | 85 | ||
| BC003903.1 | - | 2 - 3 | 287 | 3601446 | 85 | |
| BC057964.1 | - | 2 - 3 | 614 | 5352325 | 85 | |
| BC062201.1 | - | 2 - 3 | 616 | 4913425 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P377856 | BC003903.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P377856 | BC057964.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P377856 | BC062201.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P377856 | AK138669.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P377856 | AK154161.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P377856 | BC003903.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P377856 | BC057964.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P377856 | BC062201.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P377856 | AK138669.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P377856 | AK154161.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 452578 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 968729 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 758290 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 523333 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 597313 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 114822 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108639 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P377856 | BC003903.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P377856 | BC057964.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P377856 | BC062201.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P377856 | AK138669.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P377856 | AK154161.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P377856 | BC003903.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P377856 | BC057964.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P377856 | BC062201.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P377856 | AK138669.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P377856 | AK154161.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P377856 | BC003903.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P377856 | BC057964.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P377856 | BC062201.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P377856 | AK138669.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P377856 | AK154161.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P377856 | BC003903.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P377856 | BC057964.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P377856 | BC062201.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P377856 | AK138669.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P377856 | AK154161.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258589 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258615 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258597 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258601 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258583 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258587 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108639 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07111248 | - | - |
| Whole Mouse Genome G4122A | A_51_P377856 | BC003903.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P377856 | BC057964.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P377856 | BC062201.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P377856 | AK138669.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P377856 | AK154161.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P377856 | BC003903.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P377856 | BC057964.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P377856 | BC062201.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P377856 | AK138669.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P377856 | AK154161.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115505 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108639 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08803819 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08918668 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08417365 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08918669 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08918672 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08918681 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08918670 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08417363 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08918674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08494322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08364614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08517824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08918669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08587164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08364618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08727291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08495665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258595 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08364620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258585 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10369996 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369994 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369999 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369995 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369998 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369997 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423891_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10369993 | - | - |
| Mouse Genome 430 2.0 Array | 1423891_at | - | - |
| Mouse Genome 430 2.0 Array | 1423891_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 103140_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 103140_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000017043.mm.1 | NM_133994.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017043.mm.1 | NM_133994.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017043.mm.1 | TC1000003226.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000017044.mm.1 | NM_133994.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017044.mm.1 | NM_133994.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017044.mm.1 | TC1000003226.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423891_PM_at | - | - |
| Mouse Expression Set 430 | 1423891_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423891_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423891_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423891_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423891_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 610478 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5080286 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5080286 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005080286 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2665715 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2665715 | - | - |
| mouse-6 v1.1 (Illumina) | scl37812.5_586-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5080286 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37812.5_586-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5080286 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2665715 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2665715 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl37812.5_586-S | - | - |

Gstt3
Genomic Map