Assay Details
| Gene Symbol: Pnpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134021.2 | NP_598782.1 | 2 - 3 | 413 | 62 | |
| GenBank mRNA | AK154004.1 | - | 2 - 3 | 353 | 62 | |
| AK154443.1 | - | 2 - 3 | 174 | 62 | ||
| AK164667.1 | - | 2 - 3 | 414 | 62 | ||
| BC010785.1 | - | 2 - 3 | 352 | 4210325 | 62 | |
| BC026564.1 | - | 2 - 3 | 380 | 4971745 | 62 | |
| CK625276.1 | - | 2 - 3 | 222 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pnpo
Genomic Map