| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500002772.mm.1 |
NM_001168250.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002772.mm.1 |
NM_001168250.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002772.mm.1 |
NM_134092.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002772.mm.1 |
NM_134092.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002772.mm.1 |
TC1500000398.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500002770.mm.1 |
NM_001168250.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002770.mm.1 |
NM_001168250.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002770.mm.1 |
NM_134092.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002770.mm.1 |
NM_134092.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002770.mm.1 |
TC1500000398.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500002769.mm.1 |
NM_001168250.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002769.mm.1 |
NM_001168250.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002769.mm.1 |
NM_134092.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002769.mm.1 |
NM_134092.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500002769.mm.1 |
TC1500000398.mm.1 |
MTA TC |