Assay Details
| Gene Symbol: Mtbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168250.1 | NP_001161722.1 | 11 - 12 | 1366 | 91 | |
| NM_134092.3 | NP_598853.3 | 11 - 12 | 1366 | 91 | ||
| GenBank mRNA | AJ278508.1 | - | 11 - 12 | 1287 | 91 | |
| AK080036.1 | - | 11 - 12 | 1180 | 91 | ||
| AK088549.1 | - | 11 - 12 | 1180 | 91 | ||
| AK088652.1 | - | 1 - 2 | 794 | 91 | ||
| AK133855.1 | - | 11 - 12 | 1214 | 91 | ||
| BC005500.1 | - | 4 - 5 | 363 | 91 | ||
| BC052197.1 | - | 11 - 12 | 1366 | 6511752 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK088652.1 transcripts.
Set Membership: |
|

Mtbp
Genomic Map