Assay Details
| Gene Symbol: Mbnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134163.4 | NP_598924.1 | 4 - 5 | 665 | 73 | |
| GenBank mRNA | AB077700.1 | - | 3 - 4 | 571 | 73 | |
| AF466292.1 | - | 3 - 4 | 571 | 73 | ||
| AJ427920.1 | - | 3 - 4 | 571 | 73 | ||
| AK040522.1 | - | 3 - 4 | 419 | 73 | ||
| AK050908.1 | - | 3 - 4 | 384 | 73 | ||
| AK088971.1 | - | 4 - 5 | 678 | 73 | ||
| AK167342.1 | - | 3 - 4 | 384 | 73 | ||
| BC057157.1 | - | 4 - 5 | 903 | 5348959 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mbnl3
Genomic Map