Assay Details
| Gene Symbol: Mbnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134163.4 | NP_598924.1 | 5 - 6 | 904 | 76 | |
| GenBank mRNA | AB077700.1 | - | 4 - 5 | 810 | 76 | |
| AF466292.1 | - | 4 - 5 | 810 | 76 | ||
| AJ427920.1 | - | 4 - 5 | 810 | 76 | ||
| AK040522.1 | - | 4 - 5 | 658 | 76 | ||
| AK050908.1 | - | 4 - 5 | 623 | 76 | ||
| AK088971.1 | - | 5 - 6 | 917 | 76 | ||
| AK167342.1 | - | 4 - 5 | 623 | 76 | ||
| BC057157.1 | - | 5 - 6 | 1142 | 5348959 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mbnl3
Genomic Map