Assay Details
| Gene Symbol: Aldh7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127338.1 | NP_001120810.1 | 9 - 10 | 970 | 70 | |
| NM_138600.4 | NP_613066.2 | 7 - 8 | 776 | 70 | ||
| GenBank mRNA | AK004991.1 | - | 9 - 10 | 904 | 70 | |
| AK145873.1 | - | 7 - 8 | 719 | 70 | ||
| AK160117.1 | - | 7 - 8 | 703 | 70 | ||
| AK169195.1 | - | 7 - 8 | 719 | 70 | ||
| BC012407.1 | - | 7 - 8 | 738 | 4191228 | 70 | |
More Information
Set Membership: |
|

Aldh7a1
Genomic Map