Assay Details
| Gene Symbol: Aldh7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127338.1 | NP_001120810.1 | 10 - 11 | 1042 | 72 | |
| NM_138600.4 | NP_613066.2 | 8 - 9 | 848 | 72 | ||
| GenBank mRNA | AK004991.1 | - | 10 - 11 | 976 | 72 | |
| AK145873.1 | - | 8 - 9 | 791 | 72 | ||
| AK160117.1 | - | 8 - 9 | 775 | 72 | ||
| AK169195.1 | - | 8 - 9 | 791 | 72 | ||
| BC012407.1 | - | 8 - 9 | 810 | 4191228 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh7a1
Genomic Map