Assay Details
| Gene Symbol: Cds2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291039.1 | NP_001277968.1 | 2 - 3 | 170 | 65 | |
| NM_001291040.1 | NP_001277969.1 | 2 - 3 | 493 | 65 | ||
| NM_138651.7 | NP_619592.1 | 2 - 3 | 493 | 65 | ||
| GenBank mRNA | AK036328.1 | - | 2 - 3 | 344 | 65 | |
| AK049257.1 | - | 2 - 3 | 171 | 65 | ||
| AK050589.1 | - | 2 - 3 | 401 | 65 | ||
| AK147541.1 | - | 2 - 3 | 385 | 65 | ||
| AK166001.1 | - | 2 - 3 | 344 | 65 | ||
| AK170888.1 | - | 2 - 3 | 382 | 65 | ||
| AY159802.1 | - | 2 - 3 | 416 | 65 | ||
| BC003852.1 | - | 2 - 3 | 399 | 3600825 | 65 | |
| BC059776.1 | - | 2 - 3 | 296 | 65 | ||
| BC069879.1 | - | 2 - 3 | 372 | 30439069 | 65 | |
| CD540005.1 | - | 2 - 3 | 168 | 65 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK049257.1 transcripts.
Set Membership: |
|

Cds2
Genomic Map