Assay Details
| Gene Symbol: Fgd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF402611.1 | - | 4 - 5 | 919 | 116 | |
| AF402612.1 | - | 3 - 4 | 816 | 116 | ||
| AF402613.1 | - | 4 - 5 | 966 | 116 | ||
| AK028302.1 | - | 1 - 2 | 368 | 116 | ||
| AK034504.1 | - | 3 - 4 | 909 | 116 | ||
| AK043656.1 | - | 4 - 5 | 964 | 116 | ||
| AK054242.1 | - | 4 - 5 | 907 | 116 | ||
| AK134961.1 | - | 4 - 5 | 982 | 116 | ||
| AK149626.1 | - | 4 - 5 | 970 | 116 | ||
| BB569027.2 | - | 1 - 2 | 368 | 116 | ||
| BC129902.1 | - | 4 - 5 | 1006 | 40111637 | 116 | |
| BC129903.1 | - | 4 - 5 | 1007 | 40111639 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fgd4
Genomic Map