Assay Details
| Gene Symbol: Galnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139272.2 | NP_644678.2 | 5 - 6 | 675 | 86 | |
| GenBank mRNA | AF348968.1 | - | 5 - 6 | 543 | 86 | |
| AK046567.1 | - | 5 - 6 | 633 | 86 | ||
| AK145122.1 | - | 5 - 6 | 553 | 86 | ||
| AK149609.1 | - | 5 - 6 | 601 | 86 | ||
| AK151536.1 | - | 5 - 6 | 466 | 86 | ||
| BC007172.1 | - | 4 - 5 | 410 | 86 | ||
| BC053063.1 | - | 4 - 5 | 518 | 5702572 | 86 | |
| BC059818.1 | - | 5 - 6 | 620 | 5697789 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt2
Genomic Map