Assay Details
| Gene Symbol: Galnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139272.2 | NP_644678.2 | 11 - 12 | 1269 | 85 | |
| GenBank mRNA | AF348968.1 | - | 11 - 12 | 1137 | 85 | |
| AK046567.1 | - | 11 - 12 | 1227 | 85 | ||
| AK149609.1 | - | 11 - 12 | 1197 | 85 | ||
| AK151536.1 | - | 11 - 12 | 1060 | 85 | ||
| AK202473.1 | - | 1 - 2 | 37 | 85 | ||
| BC007172.1 | - | 10 - 11 | 1004 | 85 | ||
| BC053063.1 | - | 10 - 11 | 1112 | 5702572 | 85 | |
| BC059818.1 | - | 11 - 12 | 1214 | 5697789 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt2
Genomic Map